Cleanroom doekjes, B'Wipes, 7000 serie
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
115-1824EA 72.2 EUR
115-1824 115-1829
Cleanroom doekjes, B'Wipes, 7000 serie
Schoonmaakdoekjes Cleanroom doekjes Droge cleanroom doekjes
Microfibre, sealed-edge wipe for use in ISO 3 cleanrooms.
  • Microfibre polyester/nylon wipe, with sealed edges (165 g/m²)
  • Laundered, with sealed edges, for excellent cleanliness
  • Very fine microfibre filaments, densely knitted, greatly increase the quantity of contaminants picked up by the wipe, even when dry
  • Reduces the risk of scratching sensitive surfaces, such as optics
  • Ideal for cleaning sensitive surfaces, or when wet wiping is not an option