Avantor® Prevail Cyano, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH99243EA 1140 EUR
HICH99243 HICH99247 99131/N
Avantor® Prevail Cyano, HPLC Columns
Kolommen voor chromatografie
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
These general purpose cyano columns are suitable for normal or reversed phase use. They can be used for rugged normal phase applications.
Informatie over toebehoren: Use guard cartridges with holder (80101/N) and column coupler (HI-081).