Thermoelectric flanging tools, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F709-50 F709-52 F709-56 F701-12 F709-54 F701-14
BOHLF709-50EA 94.3 EUR
BOHLF709-50 BOHLF709-52 BOHLF709-56 BOHLF701-12 BOHLF709-54 BOHLF701-14
Thermoelectric flanging tools, BOLA
Gereedschapset
Thermoelectric flanging tools are used for making flanges at the ends of plastic tubing such as PTFE, PFA and FEP tubings.
Leveringsinformatie: Supplied with basic flanging tool (230 V), exchangeable flanging tip for Int.Ø 0,8 or 1,6 mm and tubing holder for tubing Ext.Ø 1,6 and 3,2 mm.