Cooling box, electrical, Powerbox® Plus
Leverancier: CAMPING GAZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2000037453 2000037452
CAMP2000037453EA 124 EUR
CAMP2000037453 CAMP2000037452
Cooling box, electrical, Powerbox® Plus
Dozen Koelboxen en koeltassen
Electrical cooling box equipped with a dial wheel, with which the power and efficiency of the box can be regulated helps to keeps the contents fresh and protected from odour, mold and fungi.
  • Premium PU insulated lid for even better performance
  • It can be used both on 12 V (auto) and 230 V (normal) socket outlet
  • Suitable for longer camping trips or outings
  • Bail handle for comfortable carrying
Antimicrobial effect is obtained from an additive which is melted down into the plastic before it is molded into the liner.