Powder bottles, PP, round, with screw top, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8890P-250S 8890P-250 8890P-500 8890P-500S
SAMP8890P-250SEA 219 EUR
SAMP8890P-250S SAMP8890P-250 SAMP8890P-500 SAMP8890P-500S
Powder bottles, PP, round, with screw top, SteriWare®
Flessen Kunststof flessen
Made of PP.
  • Moulded and packed in a cleanroom
  • FDA and EU conforming materials
  • BSE/TSE-free
  • Full batch traceability
  • Available as sterile (gamma irradiated)
These disposable powder bottles are a new premium screw top bottles that are moulded in a class 100000 cleanroom. They are designed to meet the requirements of the pharmaceutical industry.
Verpakking: Individually bagged.