Syringes for HPLC autosamplers, Gastight® CTC PAL®
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
203077 203079 203081 203083 203085 203194 203235 203274 203349 67450-01 67442-01 67444-01 67446-01 67448-01 67452-01
549-1320EA 137 EUR
549-1320 549-1322 549-1330 549-1331 549-1332 549-1318 549-1321 548-1046 549-1323 HAMI67450-01 HAMI67442-01 HAMI67444-01 HAMI67446-01 HAMI67448-01 HAMI67452-01
Syringes for HPLC autosamplers, Gastight® CTC PAL®
Spuiten Injectiespuiten voor chromatografie
CTC PAL® autosamplers are able to sample from many different vial and well types and have ultimate platform flexibility. The syringe is the most important part of every injection that the HPLC PAL® system performs. These autosampler syringes are specially designed for use in these HPLC PAL® systems: PAL® HTC-xt, PAL® HTS-xt, PAL® HTX-xt sold under the following brands: AB Sciex, Agilent, Alpha M.O.S., Antek, Bruker, Dionex, GE, Gerstel, GL Sciences, Lauda, LEAP Technologies, MicroCal, MPS, Perichrom, PerkinElmer, Shimadzu, Sotax, Thermo Scientific, Waters and Zoex.
C-Line syringesMore Product Information
Order Now
Specificaties
Environmentally Preferable