PEEK unions
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-ZRU21FPK JR-55024-5 JR-55025-5 JR-55023-5 JR-55022-5 JR-55021-5 JR-55020-5 JR-1061 JR-1066 JR-1067 JR-067 JR-066 JR-061 JR-ZN2PK-5 JR-ZBU1MFPK JR-ZBU1FPK JR-ZBU1CFPK JR-ZF.5PK-5
554-1529EA 0 EUR
554-1529 JOURJR-55024-5 JOURJR-55025-5 JOURJR-55023-5 JOURJR-55022-5 554-1512 JOURJR-55020-5 554-1505 554-1506 554-1507 554-1504 554-1503 554-1502 554-1527 554-1521 554-1520 554-1519 JOURJR-ZF.5PK-5
PEEK unions
Fittingen en connectoren
VICI Jour PEEK unions are usable with most common HPLC solvents (organic and inorganic) without the risk of corrosion, metal contamination, or protein degradation that might occur with stainless steel. Connections to these PEEK unions can be made with any 10 to 32 fitting that uses a polymeric ferrule. The unions use the standardized Valco fitting details with smooth transition of the taper into the threaded part.
  • Easy connection of transfer/solvent lines
  • Low dead volume
  • Completely inert and biocompatible
Unions are available complete with our PEEK one-piece fingertight fittings ready to connect to any 1/16" Ø tubing (PEEK, stainless steel, PTFE, ETFE, etc.), or in a 'body only' version. The body only version allows you to use colour-coded PEEK one-piece fittings for easy solvent line identifcation.
Pressure rating: <350 bar (5000 psi)