Stainless steel unions
Leverancier: VICI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
554-1526EA 58.3 EUR
554-1526 554-1525 554-1528 554-1522 554-1533 554-1532 554-1524 554-1531 554-1530
Stainless steel unions
Fittingen en connectoren
VICI Stainless Steel Unions are made for joining two pieces of 1/16" OD tubing, or an 1/8" tube. They are precision machined with a focus on the concentricity of the bore as well as on the finish of the surfaces to give an absolute leak-free connection. These Unions have internal threads, which we recommend for most applications. Internal threads make stronger connections and internal fitting details provide the lowest dead volume for high performance instrumentation.
  • Easy connection of solvent lines
  • Low dead volume
Leveringsinformatie: Unions are sold complete with all the necessary fittings. Nuts are SS303; ferrules are SS316; body is SS316.