One-piece fingertight column couplers
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-26502 JR-26503 JR-26501 JR-26504
554-1509EA 84.1 EUR
554-1509 554-1510 554-1508 554-1511
One-piece fingertight column couplers
Fittingen en connectoren
Biocompatible PEEK construction. The unique feature of the VICI Jour® column coupler is that it adjusts to fit all 10-32 fitting types. No matter what the fitting type and pilot length – Valco®, Waters®, Upchurch®, Rheodyne®, etc., the coupler adapts to bottom out in the pilot automatically, consequently creating only small void volumes.
  • Zero dead volume connections
  • Self-adjusts to fit all column designs
The column coupler is fingertight to 350 bar (5000 psi).