3,3'-Diaminobenzidine solutions kit (Bright-DAB) for immunohistochemistry
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine
BS04-110 BS04-500 BS04-999 S/BS04-20
BS04-110EA 634 EUR
BS04-110 BS04-500 BS04-999 S/BS04-20
3,3'-Diaminobenzidine solutions kit (Bright-DAB) for immunohistochemistry
3,3'-Diaminobenzidine
3,3’Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. When in the presence of peroxidase enzyme, DAB produces a brown precipitate that is insoluble in alcohol and xylene. This product comes in a two-component system consisting of a liquid stable DAB chromogen and DAB substrate buffer.
  • High-quality substrate for IHC applications.
  • Kit Components:
  • DAB Solution A: Ready–to-use Buffered H₂O₂
  • DAB Solution B: concentrated DAB solution
Bright DAB is a very stable and superior formulation of DAB. In some cases, antibodies titers may increase titers by two-fold. Bright DAB can be used both manually and on automated stainers
Certificaten: Meets CE/IVD standards
Leveringsinformatie: The Immunologic products will be shipped with blue ice. Store the products between 2°C - 8°C
Opgelet: DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. DAB can be deactivated with equal parts of Clorox.
Formule: C₁₂H₁₄N₄
MDL Nummer: MFCD00007725
CAS nummer: 91-95-2