Fittings for UHPLC, MARVELX™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UPFP-6025350 UPFP-6100150 UPFP-6100350 UPFP-6025150 UPFP-6100070
UPCHUPFP-6025350EA 379 EUR
UPCHUPFP-6025350 UPCHUPFP-6100150 UPCHUPFP-6100350 UPCHUPFP-6025150 UPCHUPFP-6100070
Fittings for UHPLC, MARVELX™, Upchurch Scientific®
Fittingen en connectoren
MarvelX biocompatible PEEK-lined stainless steel UHPLC connection system is easy to use and eliminates dead volume. It also provides higher repeat uses without sacrificing from consistent performance.
  • Reusable up to 200 times
  • Biocompatible and zero dead volume
  • Compatible with 10 to 32 coned receiving ports
  • High plate efficiency
  • Pressure rating: 1310 bar
Built with a convenient, removable fitting and changeable precision-cut flexible tubing.
Tubing type: ¹/₃₂" Ext.Ø flexible 316 stainless steel with ¹/₁₆" Ext.Ø rigid tube.