Crucibles, cylindrical, flat base, zirconium oxide
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41274.KF 41275.KF 41276.KF
41274.KFEA 454 EUR
41274.KF 41275.KF 41276.KF
Crucibles, cylindrical, flat base, zirconium oxide
Kroezen
These crucibles are manufactured from yttria stabilised zirconium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • Increased working temperature characteristics
  • Offer useful alternatives to aluminium