Cleanroom tables, ergo
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WX1201 WX1203 WX1401 WX1403 WX0604
132-0302EA 2430 EUR
132-0302 132-0314 132-0304 132-0316 PALBWX0604
Cleanroom tables, ergo
Meubilair Tafels
These cleanroom ergo tables are manufactured with 304 stainless steel with electropolished finish and Al anodized cleanroom compatible connectors.
  • Rounded front table tops
  • Excellent design conforms to highest cleanroom specifications
  • Ergonomic table legs designed to prevent operators 'knocking knees' on table
  • Height adjustable feet or casters
  • All accessories attachable
The bases are fabricated from tubing for easy cleaning and optimal airflow. The tops are designed to reduce vibrations and are very strong holding up to 80 kg.
Leveringsinformatie: More Product Information