ESD safety ankle boots, lace-up, uvex 2, vibram®, 6529
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6529.2/49 6529.2/50 6529.2/35 6529.2/36 6529.2/37 6529.2/38 6529.2/45 6529.2/46 6529.2/43 6529.2/44 6529.2/39 6529.2/40 6529.2/41 6529.2/42 6529.2/47 6529.2/48 6529.2/51 6529.2/52
114-8127EA 140 EUR
114-8127 114-8128 114-8113 114-8114- 114-8115 114-8116 114-8123 114-8124 114-8121 114-8122 114-8117 114-8118 114-8119 114-8120 114-8125 114-8126 114-8129 114-8130
ESD safety ankle boots, lace-up, uvex 2, vibram®, 6529
Schoenen en laarzen
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent high-tech microvelour upper, soft padded tongue and collar, and a dual-density slip-resistant VIBRAM® rubber sole resistant to chemicals, oils and gasoline.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction to eliminate pressure points
  • Lightweight two-layer sole with excellent shock absorption outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuffcap for additional protection
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including the chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Certificaten: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, HI, HRO, SRC