Inklapbaar vergrendelingssysteem voor schuifafsluiters
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
148647 148648 148646
118-0530EA 102.21 EUR
118-0530 BRDY148648 BRDY148646
Inklapbaar vergrendelingssysteem voor schuifafsluiters
Veiligheidssloten
This collapsible gate valve lockout is a durable, easy to store device for a wide range of gate valves. The covers easily rotate around valve handles to the fully engaged position, while also folding into smaller sizes for easy storage on maintenance carts and in lockout kits.
  • Unique knockout centre accommodates OS & Y or 'rising stem' valves and accepts locks with shackles up to 9,53 mm diameter
  • Personal padlocks and lockout tags can be applied to ensure employee safety
Max. beugeldiameter: 9,53 mm
Min. beugelopening (mm): 12,70 mm
Order Now
Specificaties
Environmentally Preferable