Oil bath attachment for IKAMAG® and IKATHERM®
Leverancier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2829500 2829400
442-0582EA 310 EUR
442-0582 442-0581
Oil bath attachment for IKAMAG® and IKATHERM®
Baden Oliebaden
The oil bath attachment H29 and H30 can be used as oil baths together with an IKAMAG® magnetic stirrer with heating or with an IKATHERM® heating plate having a diameter of 135 mm.
  • Easy cleaning and hollow cylindrical
  • Made of aluminum cast
  • Safety grip protects from bath attachment burns
  • Positioning border prevents sliding on the heating plate