Conductivity and TDS standards, natural water
Leverancier: Hach
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2974826
2974849
2974549
2974649
HACH2974826EA
15.7
EUR
HACH2974826
HACH2974849
HACH2974549
HACH2974649
Conductivity and TDS standards, natural water
Standaarden
Geleidbaarheidsstandaarden
Natural Water Standard Solution:40% sodium sulphate/40% sodium bicarbonate/20% sodium chloride.