Gas Chromatography 3-Port SilFlow Device
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
29063466 29063467 290SF302 60201-390 60201-394 60201-398 60201-401
555-9044EA 93.8 EUR
555-9044 555-9043 555-9146 HYPE60201-390 HYPE60201-394 HYPE60201-398 HYPE60201-401
Gas Chromatography 3-Port SilFlow Device
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This SilFlow 3-port device allows column switching in GC-MS without the need to vent.
  • Utilises a make up gas module and a microfluidic device
Leveringsinformatie: This device includes a gas module with microfluidics and SilTite™ FingerTite fittings for easy set up and a reliable seal.