Membrane filters, cellulose nitrate
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13004-80-ALN 11301-80-ALN 11404-80-ALN
SART13004-80-ALNEA 471 EUR
SART13004-80-ALN 512-2891 SART11404-80-ALN
Membrane filters, cellulose nitrate
Filters Membraanfilters
These cellulose nitrate membranes are suitable for air monitoring.
Verpakking: Presterilised in bags of five.
Order Now
Specificaties
Environmentally Preferable