Breaker lockout kit
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
051265 051264 090891 090892 090893
BRDY051265EA 38.61 EUR
BRDY051265 BRDY051264 118-0520 BRDY090892 BRDY090893
Breaker lockout kit
Veiligheidssloten
These breaker blockers are ideal for locking out oversized and irregularly shaped switches.
  • Self-adhesive backed rails are permanently installed onto an electrical panel; no drilling required
  • Red blocking bar indicates locked off, green bar indicates locked on
  • Use with padlock to prevent blocker from being removed
Max. switch width: 150,00 mm
Max. shackle diameter: 6,5 mm
Min. shackle clearance: 20,00 mm
Leveringsinformatie: Kit includes 2 yellow mounting rails, 1 red blocking bar, 1 green blocking bar and 3 alcohol wipes.
Opgelet: Please note:More Product Information
Order Now
Specificaties
Environmentally Preferable