Screwthread tubes, attachable
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.182.014 0.182.018 0.182.045
201-1601EA 1.9 EUR
201-1601 201-1602 201-1608
Screwthread tubes, attachable
Adapters Adapters, Buizen met schroefdraad
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.