Spout PROFI with wide thread
Leverancier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
550100 550105
HUEN550100EA 178 EUR
HUEN550100 HUEN550105
Spout PROFI with wide thread
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Made of fracture-proof and food-safe polyethylene with sleeve connection for a hose.
  • Quick liquid transfer without gurgling thanks to integrated ventilation
  • Rotatable through 360° in any position desired
  • Pressure resistance up to 0,4 bar
  • Resistant to temperature:–20 to +70 °C
  • Run-out speed >3,5 L/min
  • Connection area outside diameter = 19 mm
  • with blue knob and very resistant to acids, alkalis and other chemicals
Spigots PROFI with wide thread 22 mm for controlled emptying of liquid from barrels and jerrycans, natural-coloured.
Depth: 100 mm
Weight: 0,05 kg