Syringe tips, Ecostep™, for Stepper™ 411
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
316.010 316.050 316.500 316.010.9 316.050.9 316.500.9 316.900 316.900.9
613-4822EA 148 EUR
613-4822 613-4824 613-4826 SOCO316.010.9 SOCO316.050.9 SOCO316.500.9 SOCO316.900 SOCO316.900.9
Syringe tips, Ecostep™, for Stepper™ 411
Pipettips
These syringe tips are designed for use with the repetitive pipette Stepper™ 411. Three sizes cover the full volume range from 10 to 5000 µl.
  • Colour coding on syringe and selector button eliminates any setting error
  • Graduation helps monitoring syringe content
  • Quick insertion and removal from Stepper™
  • Made from PE / PP for excellent chemical resistance
  • Steril bioproof™ versions have a high biological purity grade: Each batch is free from human DNA, DNase, RNase and pyrogen (endotoxin)
Order Now
Specificaties
Environmentally Preferable