HPLC columns, Allure® silica (USP L3)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9160512 9160515 9160521 9160522 9160523 9160525 9160531 9160532 9160533 9160535 9160551 9160552 9160555 9160561 9160562 9160563 9160565 9160571 9160572 9160573 9160575 916050210 916050212 916050220 916050222 9160510 9160517 9160518 9160519 9160550 9160557 9160558 9160559 9160560 9160567 9160568 9160569 9160570 9160577 9160579 916051E 916052E 916053E 916055E 916056E 916057E
REST9160512EA 626 EUR
REST9160512 REST9160515 REST9160521 REST9160522 REST9160523 REST9160525 REST9160531 REST9160532 REST9160533 REST9160535 REST9160551 REST9160552 REST9160555 REST9160561 REST9160562 REST9160563 REST9160565 REST9160571 REST9160572 REST9160573 REST9160575 REST916050210 REST916050212 REST916050220 REST916050222 REST9160510 REST9160517 REST9160518 REST9160519 REST9160550 REST9160557 REST9160558 REST9160559 REST9160560 REST9160567 REST9160568 REST9160569 REST9160570 REST9160577 REST9160579 REST916051E REST916052E REST916053E REST916055E REST916056E REST916057E
HPLC columns, Allure® silica (USP L3)
Kolommen voor chromatografie
Allure® Silica HPLC columns contain a highly retentive phase for normal phase separations.
  • Very high surface area
  • Type B silica packing
Similar phases: Maxsil Si (irregularly shaped particle), LiChrospher Si-60 (irregularly shaped particle).