Syringe filters, Sartobind® IEX MA
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
93IEXQ42GB-12A 93IEXS42DB-12V 93IEXS42GB-12A 93IEXD42DB-12V 93IEXQ42DB-12V
516-1182EA 178 EUR
516-1182 516-1183 516-1184 516-1180 516-1181
Syringe filters, Sartobind® IEX MA
Filters Spuitfilters
Membrane adsorber units with ion exchange membranes (stabilised reinforced cellulose) are ideal for capturing of large proteins and viruses as no size exclusion effects can be observed.
  • Ready-to-use units
  • Reusable for more than 1000 cycles
  • High dynamic binding capacity at high flow rates
Units can be operated also with peristaltic pumps or liquid chromatography systems. Units can be connected to each other through Luer lock to increase binding capacity.
Max. operating pressure: 6 bar