Coal fly ash - constituents
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NIST-2689 NIST-2690 NIST-2691
PROONIST-2689EA 0 EUR
PROONIST-2689 PROONIST-2690 PROONIST-2691
Coal fly ash - constituents
Standaarden Standaarden voor de petroleumindustrie
These reference standard is intended for use in the evaluation of analytical methods used for the classification of coal fly ash and for the determination of constituent elements in coal fly ash or materials of a similar matrix.
A unit consists of three 10 g hermetically sealed glass vials of fly ash pulverised to less than 145 µm particle size and blended to a high degree of homogeneity.
Certificaten: Traceable to NIST and SRM.