Ginkgo biloba (extract), NIST-3247
Leverancier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROONIST-3247EA 0 EUR
PROONIST-3247
Ginkgo biloba (extract), NIST-3247
Standaarden Organische standaarden
This standard reference material is intended primarily for use in validating analytical methods for the determination of flavonoids, terpene lactones and toxic elements in ginkgo biloba extracts and similar matrices. It can be also used for quality assurance when assigning values to in-house control materials.
A unit consists of five bottles, each containing approximately 1 g of extract.
Certificaten: Traceable to NIST and SRM numbers.