Quantus® Comfort, Cleanroom Hood
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QNTS031-2CMWH00SIDEA 0 EUR
QNTS031-2CMWH00SID QNTS031-2CMWH00MID QNTS031-2CMWH00LID QNTS031-2CMWH0XLID
Quantus® Comfort, Cleanroom Hood
Werkkleding
This flat hood is made from Quantus® Comfort fabric which is a high quality polyester fabric (100% polyester weave with conductive yarn) designed for critical cleanroom applications. The fabric provides a very high wearing comfort and at the same time a great barrier against particles (98% for airborne particles >0,3 μm).
  • Flat head design with shoulder protection
  • Spiral zipper at front (plastic)
  • Optimal fit thanks to adjustable snap fastener in the back
  • Comfortable net insert at ear positions
  • Press studs to attach a face mask to the inner side of the hood; with two press studs on each side for height adjustment
  • RFID chip included
  • Autoclavable at approx. 134 °C/5 min.
Fabric weight: 100,7 g/m²
Head circumference: 57 to 65 cm, depending on size (S: 57 - 58 cm, M: 59 - 60 cm, L: 61 - 63 cm, XL: 64 - 65 cm)
Certificaten: Oeko-Tex® Standard 100 certified.