Sterilised resealable storage bag, Zyto-Sterilset® DV
Leverancier: KWP KUNSTSTOFF WERK PLUR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.100.080 2.100.081
129-1209EA 42.9 EUR
129-1209 KWPA2.100.081
Sterilised resealable storage bag, Zyto-Sterilset® DV
Zakken Opbergzakken
Sterilized LDPE cytostatic protective packaging for transport, storage, patient transfer and internal sterile transport.
  • Safe storage and transport, safe removal and reclosure by means of a pressure seal
  • Transparent
  • Bag-in-bag-Clean Room Packaging
  • Allows cytostatic drug manufacturers to safely handle sterile cytostatic preparations
Applications: Clean room and sterile pharmacy, cytostatics production, central cytostatic drug supply.
Certificaten: GMP-compliant infeed through bag-in-bag system.