Crucibles, porcelain
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5665 5666 5667 5668
USBE5665EA 6.3 EUR
USBE5665 USBE5666 USBE5667 USBE5668
Crucibles, porcelain
Kroezen
These crucibles are made from glazed porcelain.
  • Temperature resistant up to 1000 °C
Certificaten: DIN 12904