Volumetric flasks, class A, Duran glass, NS plastic stopper
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E-1572 E-1571 E-1570
228-5154EA 54.3 EUR
228-5154 228-5152 228-5150
Volumetric flasks, class A, Duran glass, NS plastic stopper
Kolven Maatkolven
Volumetric flasks are made of borosilicate glass with NS plastic (PE) stopper.
  • With blue markings
  • With standard ground joint
  • Good chemical and temperature resistance
Certificaten: Class A; DIN 12 664; ISO 1042