Inoculation loops and needles, disposable, BD Difco™
Leverancier: DIFCO BD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
220217 220214 220216 220218 220215
DIFC220217EA 180 EUR
DIFC220217 DIFC220214 DIFC220216 DIFC220218 DIFC220215
Inoculation loops and needles, disposable, BD Difco™
Uitstrijkspatels Entogen en -naalden
These sterile, disposable inoculating loops and needles made of plastic resin are smooth and flexible to facilitate uniform streaking without damaging the agar surface. They are also colour coded for easy identification.