Test sieves, 203×50 mm
Leverancier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.349.000020 60.349.000025 60.349.000032 60.349.000036 60.349.000038 60.349.000040 60.349.000045 60.349.000050 60.349.000053 60.349.000056 60.349.000063 60.349.000071 60.349.000075 60.349.000080 60.349.000090 60.349.000100 60.349.000106 60.349.000112 60.349.000125 60.349.000140 60.349.000150 60.349.000160 60.349.000180 60.349.000200 60.349.000212 60.349.000224 60.349.000250 60.349.000280 60.349.000300 60.349.000315 60.349.000355 60.349.000400 60.349.000425 60.349.000450 60.349.000500 60.349.000560 60.349.000600 60.349.000630 60.349.000710 60.349.000800 60.349.000850 60.349.000900
510-6631EA 938 EUR
510-6631 510-6632 510-6633 510-6634 510-6635 510-6636 510-6637 510-6638 510-6639 510-6640 510-6641 510-6642 510-6643 510-6644 510-6645 510-6646 510-6647 510-6648 510-6649 510-6650 510-6651 510-6652 510-6653 510-6654 510-6655 510-6656 510-6657 510-6658 510-6659 510-6660 510-6661 510-6662 510-6663 510-6664 510-6665 510-6666 510-6667 510-6668 510-6669 510-6670 510-6671 510-6672
Test sieves, 203×50 mm
Zeven Testzeven
Sieves made from 304 stainless steel.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieves are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.
Certificaten: ISO 3310-1:2001-09