DNA isolation, Genomic DNA isolation kit, Plant II Midi/Maxi, NucleoSpin®
Leverancier: MACHEREY-NAGELTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
NucleoSpin® Plant II Midi/Maxi kits are designed for the isolation of genomic DNA from plant material and fungi. Plant II Midi allows processing of up to 400 mg starting material and Plant II Maxi up to 1500 mg.
Depending on the individual sample, typical yields are in the range from 10 up to 100 μg DNA (Midi) and 50 up to 300 µg (Maxi). The eluted DNA is ready to use for southern blotting, PCR, restriction analysis.
Leveringsinformatie: Kit contains either Plant II Midi columns with collection tubes, filters (Midi), collection tubes (15 ml), buffers, RNase A; or Plant II Maxi columns with collection tubes, filters (Maxi), collection tubes (50 ml), buffers, RNase A.