IHC PAP pens
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADI9502320001 ADI9502330001
ENZOADI9502320001EA 107 EUR
ENZOADI9502320001 ENZOADI9502330001
IHC PAP pens
Kleurstoffen Microscopy and staining reagents
The IHC PAP pen provides a hydrophobic barrier when a circle is drawn around a specimen on a glass slide.
The IHC PAP (mini) pen is especially useful when working with multiple sections on a single slide because its tip is smaller than IHC PAP pen.