Avantor® Alltima, HPLC Columns, Silica
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH43884EA 0 EUR
HICH43884 HICH81405 HICH81409 HICH43877 HICH88123 HICH88171 HICH88172 HICH81116 HICH88327 96083/N 96450/N
Avantor® Alltima, HPLC Columns, Silica
Kolommen voor chromatografie
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L3
Highly polar phase. Recommended for general purpose normal phase applications.
Order Now
Specificaties
Environmentally Preferable
Oppervlakte 340 m²/g
USP code L3