Spill containment pop-up pools
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28319 28321 28323 28325
119-0312EA 121 EUR
119-0312 119-0313 119-0314 119-0315
Spill containment pop-up pools
Absorptiematerialen en bescherming tegen morsen
The space-saving and lightweight pools are designed to collect spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery. The durable PVC coated vinyl fabric is 20% heavier than other models, providing increased abrasion resistance and reliable seam adhesions.
  • Folds tightly and stows easily
  • Yellow colour provides visibility and easy identification
  • Embedded foam ring top raises containment wall as fluid level increases
Certificaten: Helps to comply with EPA for containment and spill prevention (SPCC).