Drain covers, water-filled
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28468 28470 28472 28474
119-0354EA 131 EUR
119-0354 119-0355 119-0356 119-0357
Drain covers, water-filled
Absorptiematerialen en bescherming tegen morsen
These water filled drain covers ensure regulatory compliance and prevent run-off pollutants from entering stormwater drains.
  • Easy open push plug for filling with standard water hose
  • Deploy cover as a preventative measure to seal drains before washdowns, maintenance, or other clean-up tasks that may allow contaminants into nearby water systems
  • Long lasting durable modified vinyl construction resists damage caused by UV rays
Certificaten: Helps to comply with EPA for stormwater regulations
Bestelinformatie: Order a cover size that overlaps drain opening at least 152 mm on each edge.
Opgelet: To prevent edges from losing contact with ground surface, do not fill over 127 mm in height.