Culture media, in Petri dishes, Microsart® @media
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14313-47-ACN 14314-47-ACN 14322-47-ACN
SART14313-47-ACNEA 483 EUR
SART14313-47-ACN SART14314-47-ACN SART14322-47-ACN
Culture media, in Petri dishes, Microsart® @media
Media voor microbiologie Geprepareerde media voor microbiologie
Microsart® @media are agar media dishes for microbial limit testing. They are pre-filled with different agar media types, and ready to use together with Microsart® @filter filtration units.
  • Easy to use system
  • Safe and reliable
Microsart® @media features an innovative patented lid. This active lid enables touch-free transfer of the membrane, without using any tweezers, reducing the risk of secondary contamination. In addition, the convenient, liftable interior lid provides easy access to select colonies after incubation for further analysis.