Containment berms, QuickBerm® Lite
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28556 28564 28568
119-0373EA 652 EUR
119-0373 119-0374 119-0375
Containment berms, QuickBerm® Lite
Absorptiematerialen en bescherming tegen morsen
Portable, versatile containment berm deploys quickly for emergency spill response.
  • Leakproof, tough Durable EnGuard™I, PVC coated fabric provides broad UV and chemical resistance that withstands repeated usage
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment.
Certificaten: Helps to comply with EPA for containment and spill prevention (SPCC).