Spill trays, EcoPolyBlend™
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28715 28716 28717 28718 28719
119-0392EA 102 EUR
119-0392 119-0393 119-0394 119-0395 119-0396
Spill trays, EcoPolyBlend™
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Eco-friendly spill trays are made from 100% post-industrial recycled PE will not rust or corrode.
  • Lightweight, low profile design for small-scale spill control
  • Raised rib bottom adds rigidity and keeps contents out of spills
  • Rated for service temperatures between –34 to +49 °C
Perfect for pouring and filling applications. Ideal for containing leaks from small equipment or batteries.