Diverters for ceiling leaks
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28300 28302 28308 28310 28314
119-0298EA 89.2 EUR
119-0298 119-0299 119-0309 119-0310 119-0311
Diverters for ceiling leaks
Absorptiematerialen en bescherming tegen morsen
These ceiling leak diverters redirect roof or pipe leaks to prevent from water damages. The durable, puncture resistant and watertight PVC-coated yellow fabric is designed to channel overhead pipe and ceiling leaks to containers or floor drains.
  • Available with high strength magnets for steel rafter attachment, or grommets for bungee or zip tie attachment
  • Constructed with cinched corners for quick and easy installation
  • Simply connect to a standard garden hose to drain water safely away
Flow rate: 12,5 L/min