Spill trays, Mini-Berm Flex Tray
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28442 28444
119-0342EA 137 EUR
119-0342 119-0343
Spill trays, Mini-Berm Flex Tray
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Safe and reliable for tabletop parts cleaning and fluid transfer operations involving petroleum products and acids, the Mini-Berm Flex Tray is made from a robust EnGuard™V, modified PVC coated fabric.
  • Rigid corner supports and sturdy foam walls
  • Easy to fold and store
Certificaten: Helps to comply with EPA for containment and spill prevention (SPCC).
Leveringsinformatie: Each berm comes with a 1-in NPT drain fitting and utility strap included.