Drip pads
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28460 28458 28459 28461
119-0352EA 30 EUR
119-0352 119-0348 119-0349 119-0353
Drip pads
Absorptiematerialen en bescherming tegen morsen
Drip pads can capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable
  • Made from UV and chemical-resistant modified PVC
  • Coated mesh outer layer to trap coarse debris
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs.
Certificaten: Helps to comply with EPA for spill prevention (SPCC).