Straining baskets, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H167010000 H167030000
BELAH167010000EA 473 EUR
BELAH167010000 BELAH167030000
Straining baskets, SP Bel-Art
Manden Veiligheidsmanden
Perforated baskets made from polyethylene plastic allow rapid filling and draining, and are handy for batch rinsing and transporting labware as well as straining or filtering large particles.
  • Chemically resistant; withstand acids and alkalis
  • Service temperature up to 82 °C
  • With 2 mm perforations
  • Strap handle rises 100 mm above the rim