Human recombinant TNF alpha (from E. coli)
Leverancier: ProSci
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRSI91-006EA 336 EUR
PRSI91-006
Human recombinant TNF alpha (from E. coli)
Eiwitten en peptiden
TNF alpha is a homotrimer with a subunit molecular mass of 17 kD and plays a major role in growth regulation, differentiation, inflammation, viral replication, tumorigenesis, autoimmune diseases and in viral, bacterial, fungal, and parasitic infections. Besides inducing hemorrhagic necrosis of tumors, TNF was found to be involved in tumorigenesis, tumor metastasis, viral replication, septic shock, fever, inflammation, and autoimmune diseases including Crohn’s disease, and rheumatoid arthritis as well as graft-versus-host disease.
  • This recombinant protein can be used for biological assays. For research use only.