Mouse recombinant FTO (from E. coli)
Leverancier: ProSci
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRSI90-129EA 435 EUR
PRSI90-129
Mouse recombinant FTO (from E. coli)
Eiwitten en peptiden
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.