Human recombinant EPO non-lytic (from CHO cells)
Leverancier: ProSci
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRSI90-456EA 584 EUR
PRSI90-456
Human recombinant EPO non-lytic (from CHO cells)
Eiwitten en peptiden
Erythropoietin is the principal hormone involved in the regulation of erythropoiesis by stimulating the proliferation and differentiation of erythroid progenitor cells and the maintenance of a physiological level of circulating erythrocyte mass. It is a glycoprotein produced primarily by the kidney. The biological effects of EPO are mediated by the erythropoietin receptor (Epo R). Genetic variations in EPO is associated with susceptibility to microvascular complications of diabetes type 2 (MVCD2) (including diabetic retinopathy, diabetic nephropathy leading to end-stage renal disease and diabetic neuropathy). It is used for the treatment of anemia and misused as a performance-enhancing drug in endurance athletes.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.