Universal valve lockout, base + arm
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
065401 065400
BRDY065401EA 153.3 EUR
BRDY065401 BRDY065400
Universal valve lockout, base + arm
Veiligheidssloten
These universal lockout devices are a modular system which enables users to lock out valves of different types and sises.
  • Flexibility and security
  • Available in two different sizes
Max. shackle diameter: 9 mm
Min. shackle clearance: 20 mm