Ultimate security padlocks
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
805837 805838 805839
BRDY805837EA 213.81 EUR
BRDY805837 BRDY805838 BRDY805839
Ultimate security padlocks
Veiligheidssloten
The 7 pin disk cylinder in these padlocks gives ultimate prection against manupulation and key interchanges. The shackle and body are made of highest standard hardened alloy steel.
Shackle clearance: 27,5 mm
Shackle diameter: 11,0 mm
Shackle width: 49,5 mm
Leveringsinformatie: Supplied with two keys.